• USA Home
  • 710571 - Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)magnesium(II) dihydrate

EMAIL THIS PAGE TO A FRIEND
710571 Aldrich

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)magnesium(II) dihydrate

98%

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Manganese More...
InChI Key   WTZQUDYOTSWADB-KKUWAICFSA-L
assay   98%
mp   145-150 °C

Description

Application

Reactant for precursor for deposition of magnesium on oxide films

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?