• USA Home
  • 710628 - (11bS)-4-Chloro-4,5-dihydro-3H-dinaphtho [2,1-c:1′,2′-e]phosphepine

EMAIL THIS PAGE TO A FRIEND
710628 Aldrich

(11bS)-4-Chloro-4,5-dihydro-3H-dinaphtho [2,1-c:1′,2′-e]phosphepine

  • Empirical Formula (Hill Notation) C22H16ClP

  • Molecular Weight 346.79

Purchase

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?