• USA Home
  • 71103 - L-Aspartic acid 4-tert-butyl-1-methyl ester hydrochloride

EMAIL THIS PAGE TO A FRIEND
71103 Aldrich

L-Aspartic acid 4-tert-butyl-1-methyl ester hydrochloride

≥95.0% (TLC)

  • CAS Number 2673-19-0

  • Linear Formula (CH3)3COCOCH2CH(NH2)COOCH3 · HCl

  • Molecular Weight 239.70

  •  Beilstein Registry Number 6005591

  •  MDL number MFCD00153427

  •  PubChem Substance ID 57652272

Purchase

Properties

Related Categories Amino Acids & Derivatives, Aspartic Acid, Chemical Biology, Chemical Synthesis, Peptide Chemistry More...
InChI Key   SFYKWYAIJZEDNG-RGMNGODLSA-N
assay   ≥95.0% (TLC)

Description

Packaging

1 g in poly bottle

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?