• USA Home
  • 711888 - Boc-(S)-3-thienylglycine

EMAIL THIS PAGE TO A FRIEND
711888 Aldrich

Boc-(S)-3-thienylglycine

98%

Synonym: (S)-2-[(tert-Butoxycarbonyl)amino]-2-(3-thienyl)acetic acid

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Biology, Chemical Synthesis, Chiral Building Blocks, Glycine Derivatives,
InChI Key   VJYLMXXKPBZDHN-QMMMGPOBSA-N
assay   ≥97.5% (HPLC)
  98%
optical activity   [α]/D +111±5°, c = 1 in ethanol
storage temp.   2-8°C

Description

Packaging

250 mg in glass bottle

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?