• USA Home
  • 712000 - 1-(3-Bromophenyl)-2,2,2-trifluoroethanol

EMAIL THIS PAGE TO A FRIEND
712000 Aldrich

1-(3-Bromophenyl)-2,2,2-trifluoroethanol

Synonym: (3-Bromophenyl) trifluoromethyl carbinol

  • Empirical Formula (Hill Notation) C8H6BrF3O

  • Molecular Weight 255.03

  •  Beilstein Registry Number 2262330

Purchase

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?