• USA Home
  • 712159 - 3-O-Benzyl-1,2-O-isopropylidene-α-D-xylopentodialdo-1,4-furanose

EMAIL THIS PAGE TO A FRIEND
712159 Aldrich

3-O-Benzyl-1,2-O-isopropylidene-α-D-xylopentodialdo-1,4-furanose

Synonym: (3aR,5S,6S,6aR)-6-Benzyloxy-2,2-dimethyl-tetrahydro-furo[2,3-d][1,3]dioxole-5-carbaldehyde

  • CAS Number 23558-05-6

  • Empirical Formula (Hill Notation) C15H18O5

  • Molecular Weight 278.30

  •  Beilstein Registry Number 537506

Purchase

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?