EMAIL THIS PAGE TO A FRIEND
712574 Aldrich

NHS-PEG4-(PEG12)3

≥90% (HPLC)

Synonym: Monodisperse branched PEG-NHS-ester

  • Empirical Formula (Hill Notation) C108H206N6O52

  • Molecular Weight 2420.80

  •  MDL number MFCD16621418

  •  PubChem Substance ID 329763135

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Functionalized Oligoethylene Glycol, PEGylation
InChI Key   VOPFWBJWEJMWRQ-UHFFFAOYSA-N
assay   ≥90% (HPLC)

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?