• USA Home
  • 712779 - (S)-2-Azido-3-tert-butoxypropionic acid cyclohexylammonium salt

EMAIL THIS PAGE TO A FRIEND
712779 Aldrich

(S)-2-Azido-3-tert-butoxypropionic acid cyclohexylammonium salt

≥98% (CE)

Synonym: L-Azidoserine tert-butyl ether cyclohexylammonium salt, N3-Ser(OtBu)-OH CHA salt

  • CAS Number 1228077-87-9

  • Empirical Formula (Hill Notation) C7H13N3O3 ·C6H13N

  • Molecular Weight 286.37

  •  MDL number MFCD11223827

Purchase

Properties

Related Categories Aliphatic Azides, Amino Acid Azides/Alkynes, Amino Acids & Derivatives, Asymmetric Synthesis, Azides,
InChI Key   KUVVCFAAYOBPAV-JEDNCBNOSA-N
assay   ≥98% (CE)
optical activity   [α]/D +3.7±0.3°, c = 1 in methanol
composition   carbon, 54.0-55.0%
  nitrogen, 19.1-19.9%
storage temp.   2-8°C

Description

Packaging

100 mg in glass bottle

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?