• USA Home
  • 713880 - (1R,2R)-trans-2-(Boc-amino)-1,2-dihydro-1-naphthol

EMAIL THIS PAGE TO A FRIEND
713880 Aldrich

(1R,2R)-trans-2-(Boc-amino)-1,2-dihydro-1-naphthol

≥96.0%

  • CAS Number 904316-32-1

  • Empirical Formula (Hill Notation) C15H19NO3

  • Molecular Weight 261.32

  •  MDL number MFCD14636539

Purchase

Properties

Related Categories Amino Alcohols, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks,
InChI Key   KYJBOYBBQNSQCN-CHWSQXEVSA-N
assay   ≥96.0% (HPLC)
  ≥96.0%
optical purity   enantiomeric excess: ≥97.0%
storage temp.   2-8°C

Description

Packaging

250 mg in glass bottle

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Lautens Chiral Building Blocks

Professor Mark Lautens and his group at the University of Toronto have developed a powerful catalytic asymmetric ring opening reaction enabling the production of highly functionalized hydronaphthalen...
Dr. Daniel Weibel
Chemfiles Volume 10 Article 1
Keywords: Asymmetric synthesis, Building blocks, Catalysis, Chemfiles, Clinical, Ligands, Pharmaceutical, Type

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?