• USA Home
  • 713945 - (1S,2S)-trans-1,2,3,4-Tetrahydro-1,2-naphthalenediol

EMAIL THIS PAGE TO A FRIEND
713945 Aldrich

(1S,2S)-trans-1,2,3,4-Tetrahydro-1,2-naphthalenediol

≥96.0% (HPCE/HPLC)

Synonym: (1S,2S)-trans-1,2-Tetralindiol, (1S,2S)-1,2-Dihydroxy-1,2,3,4-tetrahydronaphthalene

  • CAS Number 23190-43-4

  • Empirical Formula (Hill Notation) C10H12O2

  • Molecular Weight 164.20

  •  Beilstein Registry Number 1911276

  •  MDL number MFCD14584490

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks, Polyols More...
InChI Key   KMQJJAOZMONGLS-UWVGGRQHSA-N
assay   ≥96.0% (HPCE/HPLC)
optical purity   enantiomeric excess: ≥97.0%
storage temp.   2-8°C

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Lautens Chiral Building Blocks

Professor Mark Lautens and his group at the University of Toronto have developed a powerful catalytic asymmetric ring opening reaction enabling the production of highly functionalized hydronaphthalen...
Dr. Daniel Weibel
Chemfiles Volume 10 Article 1
Keywords: Asymmetric synthesis, Building blocks, Catalysis, Chemfiles, Clinical, Ligands, Pharmaceutical, Type

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?