• USA Home
  • 714240 - (S)-2-(Boc-amino)-5-hexynoic acid

EMAIL THIS PAGE TO A FRIEND
714240 Aldrich

(S)-2-(Boc-amino)-5-hexynoic acid

≥96% (HPLC)

Synonym: Boc-L-homopropargyl-Gly-OH, Boc-L-homopropargylglycine, Boc-Hpg-OH

  • CAS Number 208522-16-1

  • Empirical Formula (Hill Notation) C11H17NO4

  • Molecular Weight 227.26

  •  Beilstein Registry Number 10547756

  •  MDL number MFCD11052897

  •  PubChem Substance ID 329763265

Purchase

Properties

Related Categories Amino Acid Azides/Alkynes, Chemical Biology, Chemical Synthesis, Conjugation Chemistry: Azides, Alkynes and Other Reagents, Copper Click Chemistry,
InChI Key   DPHIBCOGEUVKGB-QMMMGPOBSA-N
assay   ≥96% (HPLC)
optical activity   [α]/D -80.0±2.0°, c = 1 in DMF
composition   carbon, 57.0-59.3%
  nitrogen, 5.9-6.4%

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability


Laboratory Gloves

Cross-Coupling Guide
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Alkynes and Acetylenic Building Blocks

Aldrich Chemistry lists over 600 alkyne building blocks, including a wide array of propargyl alcohols, acetylenic boron reagents, and halogenated substrates for cross-coupling. Additionally, many of ...
Keywords: Building blocks, Coupling reactions, Cross couplings

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?