Empirical Formula (Hill Notation) Au
Molecular Weight 196.97
EC Number 231-165-9
MDL number MFCD00003436
Popular Documents: Specification Sheet (PDF)
| Related Categories | 79: Au, Au Nanorods, Bioelectronics, Biomedical Materials, Biosensing and Bioimaging, |
| form | colloidal suspension |
| concentration | 100 μg/mL in H2O |
| diam. × L | 25 nm × 75 nm |
| diameter | 25 nm |
| pH range | 3.0 - 5.0 |
| density | 1 g/mL at 25 °C |
| storage temp. | 2-8°C |
10 mL in poly bottle
Do not freeze, quality lost if frozen even once. Please keep at approximately 4 °C
Customers Also Viewed

palladium coated, 25 nm diameter, 73 nm (long), 38.5 μg/mL Pd in H2O, 50 μg/mL Au in H2O
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?