• USA Home
  • 717673 - 4-Methoxy-3-pyridineboronic acid pinacol ester

EMAIL THIS PAGE TO A FRIEND
717673 Aldrich

4-Methoxy-3-pyridineboronic acid pinacol ester

99%

Purchase

Properties

Related Categories Boronate Esters, Boronic Acids and Derivatives, Chemical Synthesis, Heteroaryl Boronate Esters, Organometallic Reagents More...
InChI Key   FVHUFNSLTSKBDN-UHFFFAOYSA-N
assay   99%
mp   168-172 °C
storage temp.   −20°C

Price and Availability


KitAlysis - Increasing Catalysis Productivity
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
Protocols & Articles

Articles

Boronic Acid Esters - Chemfiles Volume 4 Article 2

The synthesis of biaryl compounds via the Suzuki–Miyaura coupling reaction has become more commonplace now that many arylboronic acids are readily available. Several years ago, Miyaura et al. demonst...
Chemfiles Volume 4 Article 2
Keywords: Coupling reactions

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?