719048 Aldrich

Silver nanoparticle ink

<150 nm particle size (DLS), 20 wt. % (dispersion in organic solvents)

DOWNLOAD MSDS (PDF)

Synonym: SunTronic® Silver

  • Linear Formula Ag

  • Molecular Weight 107.87

  •  MDL number MFCD00003397

Properties

Related Categories 47: Ag, Inks for Printing Applications, Materials Science, Metal and Ceramic Science, Metals,
form   dispersion
concentration   in ethanol and ethanediol
  20 wt. % (dispersion in organic solvents)
volume resistivity   5-30 μΩ-cmafter annealing @ 150-300 °C
particle size   <150 nm (DLS)
surface tension   28.0-31.0 dynes/cm
viscosity   11.6-13.0 cP
density   1.22 g/mL at 25 °C
storage temp.   2-8°C

Description

General description

Made using Cabot Nanoparticles

Other Notes

Ethanol and ethanediol can be used as a flush material.

Packaging

5, 25 mL in glass bottle

Legal Information

Product of Sun Chemicals

SunTronic is a registered trademark of Sun Chemical, Inc.

Physical properties

Do not freeze, quality lost if frozen even once. Please keep at approximately 4 °C.

Price and Availability

Customers Also Viewed

Silver
576832Silver

nanopowder, <100 nm particle size, contains PVP as dispersant, 99.5% trace metals basis

Silver, dispersion

nanoparticles, 10 nm particle size (TEM), 0.02 mg/mL in aqueous buffer, contains sodium citrate as stabilizer

Silver, dispersion

nanoparticle, 50-60 wt. % in tetradecane, spec. resistivity ~2.2 μΩ-cm, for printing on ITO and glass

Silver, dispersion

nanoparticle, 50-60 wt. % in tetradecane, spec. resistivity ~2.7 μΩ-cm, for printing on polyimide films

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?