Synonym: Ferratec® Brand
CAS Number 13718-66-6
Linear Formula K2FeO4
Molecular Weight 198.04
MDL number MFCD01321363
Popular Documents: Specification Sheet (PDF)
| Related Categories | Catalysis and Inorganic Chemistry, Chemical Synthesis, Iron More... |
| assay | >90% |
| mp | 276-286 °C |
| >400 °C(lit.) |
25, 100, 500 g in glass bottle
Ferratec is a registered trademark of Ferratec, LLC
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?