• USA Home
  • 72437 - (+)-Ethyl (S)-2-hydroxy-4-cyclohexylbutyrate

EMAIL THIS PAGE TO A FRIEND
72437 Aldrich

(+)-Ethyl (S)-2-hydroxy-4-cyclohexylbutyrate

≥99.0% (sum of enantiomers, GC)

  • CAS Number 402587-14-8

  • Empirical Formula (Hill Notation) C12H22O3

  • Molecular Weight 214.30

  •  Beilstein Registry Number 8982877

  •  MDL number MFCD03095458

  •  PubChem Substance ID 24886400

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Esters, Organic Building Blocks More...
InChI Key   QBVDXURIWZBADP-NSHDSACASA-N
assay   ≥99.0% (sum of enantiomers, GC)
optical activity   [α]20/D +2.5±0.5°, c = 1% in chloroform
optical purity   enantiomeric ratio: ≥99:1 (HPLC)
refractive index   n20/D 1.462

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?