Synonym:
PS-block-PAA,
Polystyrene-
Linear Formula CH3CH(C6H5) [CH2CH(C6H5)]x [CH2CH(COOH)]y Br
Popular Documents: Specification Sheet (PDF)
| Related Categories | Block Copolymers, Diblock Copolymers, Materials Science, PS-PAA, Polymer Science, |
| mol wt | Mn 83,000 |
| average Mn 13,000 (poly(acrylic acid)) | |
| average Mn 70,000 (polystyrene) | |
| mp | 420-424 °C |
| Mw/Mn | 1.1 |
Block copolymer used for making polymeric vesicles (polymersomes) and other encapsulation applications.1,2
500 mg in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?