• USA Home
  • 729167 - EDOT carboxylic acid

EMAIL THIS PAGE TO A FRIEND
729167 Aldrich

EDOT carboxylic acid

95%

Synonym: 2,3-Dihydrothieno[3,4-b][1,4]dioxine-2-carboxylic acid

Purchase

Properties

Related Categories Materials Science, Organic and Printed Electronics, Synthetic Tools and Reagents, Thiophene Monomers and Building Blocks
InChI Key   CAHFWFCOAPQXBC-UHFFFAOYSA-N
assay   95%
mp   135-140 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Conjugated Polymers for the Engineering of Device/Tissue Interface

There is considerable interest in the development of conjugated, electronically and ionically active polymers for interfacing active biomedical devices with living tissue.1-6 Conjugated polymers offe...
David C. Martin*, Laura K. Povlich and Kathleen E. Feldman
Material Matters 2010, 5.3, 68.
Keywords: Adhesion, Applications, Company, Cycloadditions, Deposition, Magnetic resonance spectroscopy, Materials Science, Methods, Polymerization reactions, Reductions, Sequences, Solution polymerization

Development of Organic Semiconductors from Highly Ordered Oligo and Polythiophenes

Nicholas S. Colella, Lei Zhang, Alejandro L. Briseño* Polymer Science & Engineering Department, University of Massachusetts, Amherst, Massachusetts 01003 *Email: abriseno@mail.pse.umass.edu
Keywords: Absorption, Building blocks, Catalysis, Coupling reactions, Cross couplings, Crystallization, Electronics, Environmental, Grignard Reaction, Metathesis, Nanomaterials, Nucleic acid annealing, Organic electronics, Photovoltaics, Polymer science, Polymerization reactions, Semiconductor, Solar cells, Ultraviolet-Visible spectroscopy

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?