• USA Home
  • 732915 - 7-Fluorochromone-3-carboxaldehyde

EMAIL THIS PAGE TO A FRIEND
732915 Aldrich

7-Fluorochromone-3-carboxaldehyde

Synonym: 3-Formyl-7-fluorochromone, 7-Fluoro-4-oxo-4H-1-benzopyran-3-carboxaldehyde, 7-Fluoro-4-oxo-4H-chromene-3-carboxaldehyde

Purchase

Description

Packaging

250 mg in glass insert

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?