• USA Home
  • 734659 - Manganese cobalt oxide

EMAIL THIS PAGE TO A FRIEND
734659 Aldrich

Manganese cobalt oxide Green Alternative

spinel, powder, <0.5 μm particle size

Synonym: MCOA

Purchase

Properties

Related Categories Alternative Energy, Cathode Materials, Chemical Synthesis, Electrode Materials, Enabling Products,
form   powder
composition   Mn1.5Co1.5O4
greener alternative product characteristics   Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
particle size   0.10-0.50 μm
  <0.5 μm
surface area   10.0-14.0 m2/g
bulk density   1.411 g/mL

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?