• USA Home
  • 737305 - 1-Aminocyclopentane-1-carboxamide

EMAIL THIS PAGE TO A FRIEND
737305 Aldrich

1-Aminocyclopentane-1-carboxamide

95%

Purchase

Properties

Related Categories Amines, Building Blocks, C6, Chemical Synthesis, Nitrogen Compounds,
InChI Key   YGVGITVCEHRBDK-UHFFFAOYSA-N
assay   95%
mp   90-95 °C

Description

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?