• USA Home
  • 737658 - (R,R)-cis EZ-CrotylMix

EMAIL THIS PAGE TO A FRIEND
737658 Aldrich

(R,R)-cis EZ-CrotylMix

Synonym: (25:1)-(3aR,7aR)-1,3-Bis(4-bromobenzyl)-2-((Z)-but-2-enyl)-2-chlorooctahydro-1H-benzo[d][1,3,2]diazasilole:scandium(III) triflate

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Crotylation
InChI Key   PRMHXJZNOJQQCI-HHDVZCLYSA-K

Description

Application

EZ-CrotylMix: A Comprehensive Solution to the Enantioselective Crotylation of Aldehydes

Legal Information

EZ-CrotylMix is a trademark of Columbia University

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?