• USA Home
  • 737682 - (S,S)-trans EZ-CrotylMix

EMAIL THIS PAGE TO A FRIEND
737682 Aldrich

(S,S)-trans EZ-CrotylMix

Synonym: (25:1)-(3aS,7aS)-1,3-Bis(4-bromobenzyl)-2-((E)-but-2-enyl)-2-chlorooctahydro-1H-benzo[d][1,3,2]diazasilole:scandium(III) triflate

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Crotylation
InChI Key   PRMHXJZNOJQQCI-KWCDKWKPSA-K
mp   95-98 °C

Description

Application

EZ-CrotylMix: A Comprehensive Solution to the Enantioselective Crotylation of Aldehydes

Legal Information

EZ-CrotylMix is a trademark of Columbia University

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?