• USA Home
  • 74082 - Boc-α-(4-fluorobenzyl)-DL-Pro-OH

EMAIL THIS PAGE TO A FRIEND
74082 Aldrich

Boc-α-(4-fluorobenzyl)-DL-Pro-OH

≥96.0% (HPLC)

Synonym: Boc-α-(4-fluorobenzyl)-DL-proline

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Proline Derivatives, Unnatural Amino Acids & Derivatives More...
InChI Key   AWTXCFLBYUGCEW-UHFFFAOYSA-N
assay   ≥96.0% (HPLC)

Description

Packaging

500 mg in poly bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Cyclic Aryl-Substitued Amino Acid Derivatives

Aromatic a-substituted proline analogs and g-substituted pyroglutamates are valuable building blocks in both medicinal and peptide chemistry. Through Suzuki coupling technology, they can be used as s...
ChemFiles Volume 1 Article 1
Keywords: Building blocks, Peptidomimetics, Suzuki coupling

Proline Derivatives and Analogs

Proline is a non-polar proteinogenic amino acid that forms a tertiary amide when incorporated into peptides. It does not have a hydrogen on the amide group and therefore cannot act as a hydrogen bond...
Chemfiles Volume 5 Article 12
Keywords: Adhesion, Biochemistry, Catalysis, Deposition, Hydroxylations, Isomerizations, Nucleic acid hybridization, Pharmaceutical, Reductions, Substitutions, Type

Proline Derivatives

Proline is a non-polar, natural amino acid that forms a tertiary amide when incorporated into peptides. Thus, it is the only proteinogenic amino acid that does not act as a hydrogen bond donor in a p...
Matthias Junkers
Aldrich ChemFiles 2008, 8.7, 8.
Keywords: Asymmetric synthesis, Chemfiles, Pharmaceutical

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?