• USA Home
  • 743100 - H-His(Trt)-HMPB-ChemMatrix® resin

EMAIL THIS PAGE TO A FRIEND
743100 Aldrich

H-His(Trt)-HMPB-ChemMatrix® resin

extent of labeling: 0.30-0.60 mmol/g loading

Synonym: 4-{3-Methoxy-4-[(N(im)-trityl-L-histidyloxy)methyl]-phenoxy}butyric acid, polymer-bound to ChemMatrix® resin

Purchase

Properties

Related Categories ChemMatrix Resins, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Polymer Supports for SPPS,
extent of labeling   0.30-0.60 mmol/g loading
particle size   35-100 mesh (wet sieved)
storage temp.   2-8°C

Description

Legal Information

ChemMatrix is a registered trademark of Matrix Innovation, Inc.

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?