• USA Home
  • 74535 - D-(+)-Norephedrine hydrochloride

EMAIL THIS PAGE TO A FRIEND
74535 Aldrich

D-(+)-Norephedrine hydrochloride

≥98.0% (AT)

Synonym: (+)-Phenylpropanolamine hydrochloride, (1S,2R)-2-Amino-1-phenyl-1-propanol hydrochloride

  • CAS Number 40626-29-7

  • Empirical Formula (Hill Notation) C9H13NO · HCl

  • Molecular Weight 187.67

  •  Beilstein Registry Number 3913470

  •  EC Number 255-008-9

  •  MDL number MFCD00064565

  •  PubChem Substance ID 57652454

Purchase

Properties

Related Categories Alkaloid, Asymmetric Synthesis, Cell Biology, Chemical Synthesis, Chiral Auxiliaries,
InChI Key   DYWNLSQWJMTVGJ-PRCZDLBKSA-N
assay   ≥98.0% (AT)
optical activity   [α]/D +33.0±1.0°, c = 7 in H2O
mp   172-175 °C

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
RC3325000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?