Sign In to View Organizational & Contract Pricing
About This Item
Linear Formula:
EuBr2
CAS Number:
Molecular Weight:
311.77
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Skip To
assay
99.99% trace metals basis
form
powder
SMILES string
Br[Eu]Br
InChI
1S/2BrH.Eu/h2*1H;/q;;+2/p-2
InChI key
PJVPGMOCWCUQHP-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service