EMAIL THIS PAGE TO A FRIEND
75513 Aldrich

HypoGel® 200 PHB

extent of labeling: ~0.7 mmol/g loading

Synonym: O-[4-(Hydroxymethyl)phenyl]penta(oxyethylene) polymer-bound

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Polymer Supports for SPPS, Resins for Fmoc-/tBu-SPPS,
extent of labeling   ~0.7 mmol/g loading
particle size   110-150 μm
storage temp.   2-8°C

Description

Legal Information

HypoGel is a registered trademark of Rapp Polymere GmbH

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

TentaGel® and HypoGel® 4-Alkoxybenzyl Alcohol Resins

TentaGel® PHB and HypoGel® PHB resins (PHB = p-hydroxybenzyl alcohol) are excellent acid-labile supports for Fmoc-/tBuSPPS of peptide acids. Detachment of the peptide can be achieved with 95% TFA. Du...
Chemfiles Volume 3 Article 4

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?