• USA Home
  • 75540 - Tributyl orthoformate

EMAIL THIS PAGE TO A FRIEND
75540 Aldrich

Tributyl orthoformate

≥99.0% (GC)

  • CAS Number 588-43-2

  • Linear Formula CH[O(CH2)3CH3]3

  • Molecular Weight 232.36

  •  Beilstein Registry Number 1752526

  •  EC Number 209-618-7

  •  MDL number MFCD00015250

  •  PubChem Substance ID 24886841

Purchase

Properties

Related Categories Acetals/Ketals/Ortho Esters, Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds More...
InChI Key   SGJBIFUEFLWXJY-UHFFFAOYSA-N
assay   ≥99.0% (GC)
refractive index   n20/D 1.418
bp   120-122 °C/15 mmHg(lit.)
density   0.872 g/mL at 20 °C(lit.)

Description

Packaging

10 mL in glass bottle

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
192.2 °F
Flash Point(C) 
89 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?