• USA Home
  • 75609 - Osmanthus oil from Osmanthus fragrans Lour.

EMAIL THIS PAGE TO A FRIEND
75609 Aldrich

Osmanthus oil from Osmanthus fragrans Lour.

absolute, natural

Purchase

Properties

quality   natural
refractive index   n20/D 1.49
storage temp.   2-8°C

Description

Packaging

1 mL in glass bottle

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?