• USA Home
  • 756474 - (2-Di-tert-butylphosphinobiphenyl)gold(I) bis(trifluoromethanesulfonyl)imide

EMAIL THIS PAGE TO A FRIEND
756474 Aldrich

(2-Di-tert-butylphosphinobiphenyl)gold(I) bis(trifluoromethanesulfonyl)imide

97%

Synonym: (2-Biphenyl)(di-tert-butylphosphino)gold(I) bis(trifluoromethane)sulfonimide, JohnPhos AuNTf2

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Gold, Gold Catalysts
InChI Key   IXAZMTRZKJUWDK-UHFFFAOYSA-N
assay   97%
mp   207-214 °C

Description

Packaging

100, 500 mg in glass bottle

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?