• USA Home
  • 75870 - α-Oxo-2-furanacetic acid

EMAIL THIS PAGE TO A FRIEND
75870 Aldrich

α-Oxo-2-furanacetic acid

≥97.0% (T)

Synonym: (2-Furyl)glyoxylic acid

  • CAS Number 1467-70-5

  • Empirical Formula (Hill Notation) C6H4O4

  • Molecular Weight 140.09

  •  Beilstein Registry Number 115630

  •  EC Number 215-990-1

  •  MDL number MFCD00042671

  •  PubChem Substance ID 57652589

Purchase

Properties

Related Categories Building Blocks, C4 to C7, Chemical Synthesis, Furans, Heterocyclic Building Blocks More...
InChI Key   IXVPCJUAKDVYKX-UHFFFAOYSA-N
assay   ≥97.0% (T)
mp   92-97 °C

Description

Packaging

5 g in glass bottle

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGIII
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?