• USA Home
  • 76000 - Palladium on activated alumina

EMAIL THIS PAGE TO A FRIEND
76000 Aldrich

Palladium on activated alumina

10% Pd basis

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Heterogeneous Pd Catalysts, Palladium
assay   10% Pd basis

Description

Application

Hydrogenation catalyst

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?