• USA Home
  • 761354 - (Acetonitrile)(2-{bis[3,5-bis(trifluoromethyl)phenyl]phosphino}-3,6-dimethoxy-2′,4′,6′-triisopropyl-1,1′-biphenyl)gold(I) hexafluoroantimonate

EMAIL THIS PAGE TO A FRIEND
761354 Aldrich

(Acetonitrile)(2-{bis[3,5-bis(trifluoromethyl)phenyl]phosphino}-3,6-dimethoxy-2′,4′,6′-triisopropyl-1,1′-biphenyl)gold(I) hexafluoroantimonate

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Gold, Gold Complexes
InChI Key   MFKHGPCNKYFWOG-UHFFFAOYSA-H
mp   103-189 °C (decomposition)

Description

Packaging

1 g in glass bottle

250 mg in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?