EMAIL THIS PAGE TO A FRIEND
76241 Aldrich

Paraffin wax

pellets, white

Purchase

Properties

Related Categories Engineering Polymers, Materials Science, Polymer Science, Polymers, Waxes and Oils More...
form   pellets
ign. residue   ≤0.05%
color   white
mp   43-95 °C
transition temp   solidification point 50-52 °C

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
RV0350000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?