762938 Aldrich

(S)-(DTBM-SEGPHOS®)CuH

0.001 M in toluene

DOWNLOAD MSDS (PDF)

Synonym: Hydro[(4S)-[4,4′-bi-1,3-benzodioxole]-5,5′-diylbis [bis[3,5-bis(1,1-dimethylethyl)-4-methoxyphenyl]phosphine-P]]copper(I) in toluene

  • Empirical Formula (Hill Notation) C74H101CuO8P2

  • Molecular Weight 1244.08

  •  EC Number 203-625-9

Properties

Related Categories Building Blocks, C22 to C40+, Chemical Synthesis, Ethers, Organic Building Blocks,
concentration   0.001 M in toluene
density   0.879 g/mL at 25 °C
storage temp.   2-8°C

Description

Packaging

10, 50 mL in glass bottle

Application

Takasago Ligands and Complexes for Asymmetric Reactions

Legal Information

SEGPHOS® is a registered trademark of Takasago Intl. Corp.

SEGPHOS is a registered trademark of Takasago Intl. Corp.

Price and Availability

Customers Also Viewed

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?