EMAIL THIS PAGE TO A FRIEND
76535 Aldrich

2-Pentadecanone

≥98.5%

Synonym: Methyl tridecyl ketone

  • CAS Number 2345-28-0

  • Linear Formula CH3CO(CH2)12CH3

  • Molecular Weight 226.40

  •  Beilstein Registry Number 1764872

  •  EC Number 219-064-8

  •  MDL number MFCD00053712

  •  PubChem Substance ID 24887011

Purchase

Properties

Related Categories Building Blocks, C15 to C38, Carbonyl Compounds, Cell Biology, Chemical Synthesis,
InChI Key   CJPNOLIZCWDHJK-UHFFFAOYSA-N
assay   ≥98.5%
bp   293 °C(lit.)
mp   37-41 °C(lit.)
  37-41 °C

Description

Packaging

1 g in glass bottle

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?