• USA Home
  • 76634 - 2-Cyclopropyl-2-propylamine p-toluenesulfonate salt

EMAIL THIS PAGE TO A FRIEND
76634 Aldrich

2-Cyclopropyl-2-propylamine p-toluenesulfonate salt

≥98.5% (CHN)

Synonym: α,α-Dimethylcyclopropylmethylamine p-toluenesulfonate salt

  • CAS Number 172947-14-7

  • Empirical Formula (Hill Notation) C6H13N · C7H8O3S

  • Molecular Weight 271.38

  •  Beilstein Registry Number 8285125

  •  MDL number MFCD01090985

  •  PubChem Substance ID 57652652

Purchase

Properties

Related Categories Amine Salts, Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   RGWMDWPKOOWZSF-UHFFFAOYSA-N
assay   ≥98.5% (CHN)
mp   167-171 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?