• USA Home
  • 76958 - 4-(Fmoc-amino)piperidine hydrochloride

EMAIL THIS PAGE TO A FRIEND
76958 Aldrich

4-(Fmoc-amino)piperidine hydrochloride

≥95.0%

Synonym: N(4)-Fmoc-4-piperidinamine hydrochloride

  • CAS Number 221352-86-9

  • Empirical Formula (Hill Notation) C20H22N2O2 · HCl

  • Molecular Weight 358.86

  •  MDL number MFCD01321024

  •  PubChem Substance ID 57652669

Purchase

Properties

Related Categories Building Blocks, C16 to C36, Chemical Synthesis, Heterocyclic Building Blocks, Piperidines More...
InChI Key   MOAIDLUWFCXIJA-UHFFFAOYSA-N
assay   ≥95.0%

Description

Application

Combinatorial reactant for ergopeptides with high affinities for dopamine receptors

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?