• USA Home
  • 77286 - Octadecafluorooctane Fraction

EMAIL THIS PAGE TO A FRIEND
77286 Aldrich

Octadecafluorooctane Fraction

≥97.0% (GC)

Synonym: Perfluorooctane fraction

  • Linear Formula CF3(CF2)6CF3

  •  Beilstein Registry Number 1717142

  •  EC Number 206-199-2

  •  MDL number MFCD00042083

  •  PubChem Substance ID 57652693

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, F-Tagged, Fluorinated Building Blocks, Organic Fluorinated Building Blocks More...
InChI Key   YVBBRRALBYAZBM-UHFFFAOYSA-N
assay   ≥97.0% (GC)
composition   C5-, C6-, C7-homologues, ~30%
  octadecafluorooctane, ~70%
bp   103-106 °C(lit.)

Description

Packaging

50 mL in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
RG9701000
Protocols & Articles

Articles

Solvents for Fluorous Synthesis

Following the definition offered by Gladysz and Curran[1] a “Fluorous Medium” is “any phase of a perfluoroalkane, perfluorodialkylether, perfluorotrialkylamine, or similar non-polar species, or any s...
ChemFiles Volume 4 Article 9
Keywords: Methods, Separation, Solvents

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?