EMAIL THIS PAGE TO A FRIEND
77780 Aldrich

3-Phenoxytoluene

≥98.0% (GC)

Synonym: 3-Methyldiphenyl ether, Phenyl m-tolyl ether

  • CAS Number 3586-14-9

  • Linear Formula C6H5OC6H4CH3

  • Molecular Weight 184.23

  •  Beilstein Registry Number 2045714

  •  EC Number 222-716-4

  •  MDL number MFCD00008531

  •  PubChem Substance ID 57652711

Purchase

Properties

Related Categories Building Blocks, C13, Chemical Synthesis, Ethers, Organic Building Blocks,
InChI Key   UDONPJKEOAWFGI-UHFFFAOYSA-N
assay   ≥98.0% (GC)
refractive index   n20/D 1.573(lit.)
  n20/D 1.573
bp   271-273 °C(lit.)
density   1.051 g/mL at 25 °C(lit.)

Description

Packaging

100 mL in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
DA6142000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?