EMAIL THIS PAGE TO A FRIEND
778540 Aldrich

1-Iodo-1-hexyne

≥97.0% (GC)

  • CAS Number 1119-67-1

  • Linear Formula IC2(CH2)3CH3

  • Molecular Weight 208.04

  •  Beilstein Registry Number 1740148

  •  MDL number MFCD00160789

  •  PubChem Substance ID 329767978

Purchase

Properties

Related Categories Alkynes, Alkynyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons,
InChI Key   SACKEEAAIYXWOZ-UHFFFAOYSA-N
assay   ≥97.0% (GC)
density   1.564 at 20 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NA 1993 / PGIII
WGK Germany 
3
Flash Point(F) 
152.6 °F
Flash Point(C) 
67 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?