EMAIL THIS PAGE TO A FRIEND
78174 Aldrich

3-Phenylbicyclohexyl

mixture of isomers

Synonym: 1-Cyclohexyl-3-phenylcyclohexane

  • CAS Number 33460-02-5

  • Empirical Formula (Hill Notation) C18H26

  • Molecular Weight 242.40

  •  Beilstein Registry Number 2264721

  •  MDL number MFCD01863105

  •  PubChem Substance ID 24887269

Purchase

Properties

Related Categories Arenes, Building Blocks, Chemical Synthesis, Organic Building Blocks
InChI Key   ONQUBGYEUGJCEM-UHFFFAOYSA-N
composition   meta-compound percentage, 57-69%
  hydrogenation percentage, 30-35%
refractive index   n20/D 1.573
density   1.004 g/mL at 20 °C(lit.)

Description

Application

as high-heating carrier liquid

Packaging

1 L in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
228.2 °F
Flash Point(C) 
109 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?