• USA Home
  • 78205 - (±)-3-Bromo-1-phenyl-2-pyrrolidinone

EMAIL THIS PAGE TO A FRIEND
78205 Aldrich

(±)-3-Bromo-1-phenyl-2-pyrrolidinone

≥95.0% (GC)

  • CAS Number 77868-83-8

  • Empirical Formula (Hill Notation) C10H10BrNO

  • Molecular Weight 240.10

  •  Beilstein Registry Number 5004253

  •  MDL number MFCD00800296

  •  PubChem Substance ID 57652733

Purchase

Properties

Related Categories Building Blocks, C9 to C10, Chemical Synthesis, Halogenated Heterocycles, Heterocyclic Building Blocks,
InChI Key   OXRJWDQGHVJQAU-UHFFFAOYSA-N
assay   ≥95.0% (GC)
mp   100-103 °C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?