EMAIL THIS PAGE TO A FRIEND
78619 Aldrich

HypoGel® 200 HMB

extent of labeling: ~0.8 mmol/g loading

Synonym: O-[4-(Hydroxymethyl)benzamidoethyl]tetra(oxyethylene) polymer-bound

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Polymer Supports for SPPS, Resins for Fmoc-/tBu-SPPS,
extent of labeling   ~0.8 mmol/g loading
particle size   110-150 μm
storage temp.   2-8°C

Description

Legal Information

HypoGel is a registered trademark of Rapp Polymere GmbH

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Resins for the Synthesis of C-Terminal Modified Peptides

Hydroxymethylbenzoic acid (HMBA) is a useful and versatile benzyl ester derived linker, completely resistant to acids (even to liquid HF) but easily cleavable by a variety of nucleophilic reagents to...
ChemFiles Volume 3 Article 4
Keywords: Azide formation, Condensations, Peptide synthesis, Reductions, Saponifications, Solid phase peptide synthesis, Transesterifications

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?