• USA Home
  • 78918 - (S)-(−)-1-Phenyl-1,3-propanediol

EMAIL THIS PAGE TO A FRIEND
78918 Aldrich

(S)-(−)-1-Phenyl-1,3-propanediol

puriss., ≥99.0% (sum of enantiomers, GC)

  • CAS Number 96854-34-1

  • Empirical Formula (Hill Notation) C9H12O2

  • Molecular Weight 152.19

  •  Beilstein Registry Number 3603772

  •  MDL number MFCD00221510

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks, Polyols More...
InChI Key   RRVFYOSEKOTFOG-VIFPVBQESA-N
grade   puriss.
assay   ≥99.0% (sum of enantiomers, GC)
optical activity   [α]20/D −69±2°, c = 1% in chloroform stab. with amylenes
mp   61-64 °C

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?