• USA Home
  • 78974 - (S)-1-Phenyl-2-propen-1-ol

EMAIL THIS PAGE TO A FRIEND
78974 Aldrich

(S)-1-Phenyl-2-propen-1-ol

≥97.0% (sum of enantiomers, GC)

Synonym: (S)-α-Vinylbenzyl alcohol

  • CAS Number 39623-35-3

  • Empirical Formula (Hill Notation) C9H10O

  • Molecular Weight 134.18

  •  Beilstein Registry Number 5476649

  •  MDL number MFCD00210082

  •  PubChem Substance ID 24887393

Purchase

Properties

Related Categories Alcohols, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
InChI Key   MHHJQVRGRPHIMR-VIFPVBQESA-N
assay   ≥97.0% (sum of enantiomers, GC)
optical activity   [α]20/D −1.0±0.5°, c = 3.2% in chloroform
optical purity   enantiomeric ratio: ≥97:3 (GC)
refractive index   n20/D 1.543
density   1.021 g/mL at 20 °C(lit.)
storage temp.   2-8°C

Description

Packaging

500 mg in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?