• USA Home
  • 79256 - S-(4-Tolyl)-L-cysteine

EMAIL THIS PAGE TO A FRIEND
79256 Aldrich

S-(4-Tolyl)-L-cysteine

produced by Wacker Chemie AG, Burghausen, Germany, ≥95.0% (HPLC)

Synonym: (R)-2-Amino-3-(4-methylphenylthio)propionic acid, (R)-2-Amino-3-(p-tolylthio)propionic acid

  • Empirical Formula (Hill Notation) C10H13NO2S

  • Molecular Weight 211.28

  •  Beilstein Registry Number 2581850

  •  MDL number MFCD03791102

  •  PubChem Substance ID 57652782

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Cysteine Derivatives, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
grade   produced by Wacker Chemie AG, Burghausen, Germany
InChI Key   CHRJYKJYNAKSHG-VIFPVBQESA-N
assay   ≥95.0% (HPLC)
optical purity   ee: >99% (due to stereospecific enzymatic synthesis)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Articles

Unnatural Alanine and Cysteine Derivatives Obtained by Fermentation

Sigma-Aldrich offers a unique selection of unnatural amino acid derivatives in collaboration with WACKER Fine Chemicals. They have developed a fermentation process for the commercial production of th...
ChemFiles Volume 5 Article 12
Keywords: Fermentation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?