• USA Home
  • 79524 - Potassium P,P-dimethylphosphonoacetate

EMAIL THIS PAGE TO A FRIEND
79524 Aldrich

Potassium P,P-dimethylphosphonoacetate

≥95.0% (NT)

Synonym: P,P-Dimethylphosphonoacetic acid potassium salt, Phosphonoacetic acid P,P-dimethyl ester potassium salt

  • CAS Number 34170-88-2

  • Linear Formula (CH3O)2P(O)CH2COOK

  • Molecular Weight 206.18

  •  Beilstein Registry Number 6016967

  •  MDL number MFCD00042970

  •  PubChem Substance ID 57652800

Purchase

Properties

Related Categories C-C Bond Formation, Chemical Synthesis, Horner-Wadsworth-Emmons Reagents, Olefination, Synthetic Reagents More...
InChI Key   WPDZWGGYOFPQEO-UHFFFAOYSA-M
assay   ≥95.0% (NT)
mp   113-115 °C
storage temp.   2-8°C

Description

Other Notes

Starting material for preparing various mixed triesters of phosphonoacetate.; Wittig-Horner reactions of dianion

Application

Reactant for:
• Preparation of Michael acceptor-containing oligopeptide-based inhibitors
• Condensation reactions with chloroacetophenone

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?