• USA Home
  • 79898 - O-[2-(Boc-amino)-ethyl]-O′-[2-(diglycolyl-amino)ethyl]decaethylene glycol

EMAIL THIS PAGE TO A FRIEND
79898 Aldrich

O-[2-(Boc-amino)-ethyl]-O′-[2-(diglycolyl-amino)ethyl]decaethylene glycol

≥90% (oligomer purity)

Synonym: Boc-PEG-acid (n=11)

  • Empirical Formula (Hill Notation) C33H64N2O17

  • Molecular Weight 760.86

  •  MDL number MFCD03453251

  •  PubChem Substance ID 57652819

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, High Oligomer Purity PEG, PEGylation
InChI Key   FTVQYNSXSAAXQP-UHFFFAOYSA-N
assay   ≥90% (oligomer purity)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?